This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01819224_m1
See other SLO2 GE Assays ›
Gene Symbol:
SLO2
Gene Name:
slowpoke 2
Gene Aliases:
CG12897, CG12899, CG12901, CG12904, CG34363, CG42732, Dm_2R:24301, Dm_2R:24438, Dmel\CG42732, Dmel_CG42732, Slack, dSLO2
Chromosome Location:
Chr.2R: 10322195 - 10416996 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0261698
Assay ID Dm01819224_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103788.3 NP_001097258.2 9 - 10 1501 62
NM_001103789.3 NP_001097259.2 9 - 10 1501 62
NM_001201971.2 NP_001188900.1 9 - 10 1797 62
NM_001201972.3 NP_001188901.1 9 - 10 2014 62
NM_001259313.2 NP_001246242.1 9 - 10 2014 62
NM_001273932.1 NP_001260861.1 9 - 10 2014 62
NM_001273933.1 NP_001260862.1 8 - 9 1004 62
NM_001273934.1 NP_001260863.1 9 - 10 1493 62
NM_001273935.1 NP_001260864.1 9 - 10 2014 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
potassium ion transmembrane transport
intracellular sodium activated potassium channel activity
calcium-activated potassium channel activity
outward rectifier potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0