Assay Details
Gene Symbol: eIF3j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299331.1 | NP_001286260.1 | 3 - 4 | 377 | 75 | |
NM_136697.3 | NP_610541.1 | 3 - 4 | 512 | 75 | ||
GenBank mRNA | AY051457.1 | - | 3 - 4 | 357 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
