This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01816887_g1
See other ATE1 GE Assays ›
Gene Symbol:
Ate1
Gene Name:
Arginyltransferase 1
Gene Aliases:
ATE1, Ate, CG9204, Dm-Ate1, Dmel\CG9204, Dmel_CG9204, ate1, l(2)k10809
Chromosome Location:
Chr.2R: 20254966 - 20257390 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0025720
Assay ID Dm01816887_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ate1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001274172.1 NP_001261101.1 6 - 7 1321 73
NM_058046.5 NP_477394.3 6 - 7 1297 73
NM_166379.3 NP_725939.2 6 - 7 1318 73
NM_166380.3 NP_725940.2 6 - 7 1300 73
GenBank mRNA AF079101.1 - 6 - 7 1267 73
AY051688.1 - 6 - 7 1267 73
BT001498.1 - 6 - 7 1288 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

protein arginylation
ubiquitin-dependent protein catabolic process via the N-end rule pathway
arginyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0