This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01814237_g1
See other TIM GE Assays ›
Gene Symbol:
tim
Gene Name:
timeless
Gene Aliases:
CG3234, Dmel\CG3234, Dmel_CG3234, Ritsu, TIM, Tim, Tim-1, dTIM, dTim, dtim, mel_tim, rit, s-tim, tim1
Chromosome Location:
Chr.2L: 3493986 - 3508119 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0014396
Assay ID Dm01814237_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tim
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144308.3 NP_001137780.2 13 - 14 3628 83
NM_001347756.1 NP_001334730.1 12 - 13 3681 83
NM_164540.4 NP_722912.3 12 - 13 4012 83
NM_164541.6 NP_722913.3 12 - 13 3685 83
NM_164542.5 NP_722914.3 12 - 13 3681 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

DNA replication checkpoint
negative regulation of transcription from RNA polymerase II promoter
circadian regulation of heart rate
DNA repair
mating behavior
copulation
rhythmic behavior
circadian rhythm
eclosion rhythm
photoperiodism
entrainment of circadian clock
sleep
regulation of protein import into nucleus
regulation of circadian sleep/wake cycle
replication fork arrest
regulation of circadian sleep/wake cycle, sleep
locomotor rhythm
negative phototaxis
replication fork protection
circadian behavior
regulation of phagocytosis
positive regulation of phagocytosis
circadian temperature homeostasis
cellular response to light stimulus
negative regulation of transcription regulatory region DNA binding
DNA binding
protein binding
transcription factor binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0