This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01813245_g1
See other RDGA GE Assays ›
Gene Symbol:
rdgA
Gene Name:
retinal degeneration A
Gene Aliases:
CG10966, CG12660, CG15352, CG34344, CG42667, DGK, DGK2, Dmel\CG42667, Dmel_CG42667, RDGA, RdgA, dDGK2, dgk-5, x35
Chromosome Location:
Chr.X: 8907068 - 9029476 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0261549
Assay ID Dm01813245_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rdgA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042799.2 NP_001036264.2 15 - 16 3417 57
NM_001103446.2 NP_001096916.1 15 - 16 3114 57
NM_001103447.4 NP_001096917.1 16 - 17 4663 57
NM_001201635.1 NP_001188564.1 16 - 17 3471 57
NM_001201636.1 NP_001188565.1 15 - 16 5272 57
NM_001272437.1 NP_001259366.1 15 - 16 5302 57
NM_001272438.1 NP_001259367.1 15 - 16 5296 57
NM_001298098.1 NP_001285027.1 13 - 14 5186 57
NM_001298099.1 NP_001285028.1 14 - 15 5250 57
NM_078537.3 NP_511092.2 15 - 16 5287 57
GenBank mRNA AY075349.1 - 15 - 16 5290 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidic acid biosynthetic process
phosphatidylinositol biosynthetic process
protein kinase C-activating G-protein coupled receptor signaling pathway
phototransduction
sensory perception of sound
sensory perception of smell
rhodopsin mediated signaling pathway
deactivation of rhodopsin mediated signaling
intracellular signal transduction
thermotaxis
photoreceptor cell maintenance
diacylglycerol metabolic process
glycerolipid metabolic process
lipid phosphorylation
NAD+ kinase activity
diacylglycerol kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0