This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01810172_g1
See other NPC1A GE Assays ›
Gene Symbol:
Npc1a
Gene Name:
Niemann-Pick type C-1a
Gene Aliases:
CG5722, DmNPC, Dmel\CG5722, Dmel_CG5722, NPC-1, NPC1, NPC1a, Npc1, Npc1_Dm, dNPC1, dNPC1a, dmNPC1, dncp1a, dnpc1a, npc1, npc1a
Chromosome Location:
Chr.2L: 10213812 - 10221216 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0024320
Assay ID Dm01810172_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npc1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001201840.2 NP_001188769.1 5 - 6 667 128
NM_001201841.2 NP_001188770.1 5 - 6 629 128
NM_001201842.2 NP_001188771.1 5 - 6 595 128
NM_001298866.1 NP_001285795.1 5 - 6 771 128
NM_001298867.1 NP_001285796.1 5 - 6 592 128
NM_001298868.1 NP_001285797.1 5 - 6 1585 128
NM_135513.5 NP_609357.2 5 - 6 771 128

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

sperm individualization
molting cycle, chitin-based cuticle
sterol metabolic process
regulation of cholesterol transport
ecdysteroid biosynthetic process
lipid transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0