This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809523_g1
See other PAPLA1 GE Assays ›
Gene Symbol:
PAPLA1
Gene Name:
Phosphatidic Acid Phospholipase A1
Gene Aliases:
CG14278, CG8552, Dmel\CG8552, Dmel_CG8552, Sec23-IP
Chromosome Location:
Chr.2L: 8134247 - 8159638 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031990
Assay ID Dm01809523_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAPLA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001201785.1 NP_001188714.1 3 - 4 4247 69
NM_001201786.2 NP_001188715.1 3 - 4 4150 69
NM_001298812.1 NP_001285741.1 3 - 4 4437 69
NM_135341.1 NP_609185.1 3 - 4 4374 69
GenBank mRNA BT023778.1 - 3 - 4 4374 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
spermatogenesis
rhodopsin biosynthetic process
phospholipase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0