This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809244_g1
See other MEFG1 GE Assays ›
Gene Symbol:
mEFG1
Gene Name:
mitochondrial translation elongation factor G 1
Gene Aliases:
B, CG4567, Dmel\CG4567, Dmel_CG4567, G, GFM1, dEF-G1, ico, l(2)27Cb, l(2)27Ea
Chromosome Location:
Chr.2L: 7220369 - 7223081 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263133
Assay ID Dm01809244_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mEFG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_135261.2 NP_609105.1 3 - 4 2194 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial translational elongation
translation elongation factor activity
GTPase activity
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0