This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809195_g1
See other MME1 GE Assays ›
Gene Symbol:
MME1
Gene Name:
Mitochondrial Magnesium Exporter 1
Gene Aliases:
CG3476, Dmel\CG3476, Dmel_CG3476, dMme1
Chromosome Location:
Chr.2L: 7027526 - 7028959 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031881
Assay ID Dm01809195_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MME1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_135249.3 NP_609093.1 2 - 3 838 84
GenBank mRNA AY069591.1 - 2 - 3 770 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
acyl carnitine transport
cellular magnesium ion homeostasis
carnitine transport
magnesium ion transmembrane transport
magnesium ion export from mitochondrion
carnitine:acyl carnitine antiporter activity
magnesium ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0