This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01808160_g1
See other AP-2ALPHA GE Assays ›
Gene Symbol:
AP-2alpha
Gene Name:
Adaptor Protein complex 2, alpha subunit
Gene Aliases:
AP-2, AP2, AP2alpha, Ada, Alpha-Adaptin, CG31654, CG4260, D-Ada, D-alphaAda, Dmel\CG4260, Dmel_CG4260, MENE (2L)-A, MENE(2L)-A, ada, alpha, alpha-Ada, alpha-Adaptin, alpha-ada, alpha-adaptin, alphaAda, alphaada, apl3, cg4260, dAP-2a, l(2)06694, l(2)SH0460, l(2)SH2 0460
Chromosome Location:
Chr.2L: 408030 - 414777 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0264855
Assay ID Dm01808160_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AP-2alpha
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057471.6 NP_476819.2 7 - 8 3069 73
NM_205885.2 NP_995607.1 6 - 7 2636 73
GenBank mRNA BT032839.1 - 7 - 8 3050 73
BT125866.1 - 6 - 7 2636 73
Y11104.1 - 2938 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
endocytosis
receptor-mediated endocytosis
protein localization
asymmetric cell division
positive regulation of autophagy
protein transport
regulation of endocytosis
ovarian follicle cell development
positive regulation of endocytosis
synaptic vesicle endocytosis
synaptic vesicle transport
clathrin-mediated endocytosis
mitotic cleavage furrow ingression
clathrin adaptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0