This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01805337_g1
See other PLP GE Assays ›
Gene Symbol:
Plp
Gene Name:
Pericentrin-like protein
Gene Aliases:
AKAP450, CG13459, CG18648, CG33957, CG6735, CP309, Cp309, D-PLP, D-Plp, DPLP, DPlp, Dmel\CG33957, Dmel_CG33957, Dplp, PACT, PCNT, PLP, cp309, d-plp, dPLP, dplp, l(3)s2172, pPlp, plp
Chromosome Location:
Chr.3L: 15066081 - 15094724 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0086690
Assay ID Dm01805337_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001032402.2 NP_001027574.2 4 - 5 3449 57
NM_001259833.2 NP_001246762.2 5 - 6 3404 57
NM_001274930.1 NP_001261859.1 4 - 5 3290 57
NM_001274932.1 NP_001261861.1 5 - 6 3404 57
NM_001274934.1 NP_001261863.1 4 - 5 3449 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

microtubule nucleation
centrosome cycle
signal transduction
sensory perception of sound
cilium-dependent cell motility
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0