This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01803605_g1
See other PGAP5 GE Assays ›
Gene Symbol:
PGAP5
Gene Name:
Post-GPI attachment to proteins 5
Gene Aliases:
CG8455, Dmel\CG8455, Dmel_CG8455
Chromosome Location:
Chr.2L: 8198882 - 8200652 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031997
Assay ID Dm01803605_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001273302.1 NP_001260231.1 2 - 3 1091 70
NM_135347.3 NP_609191.1 2 - 3 1020 70
NM_164798.3 NP_723359.1 2 - 3 1020 70
GenBank mRNA AY094655.1 - 2 - 3 935 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor biosynthetic process
ER to Golgi vesicle-mediated transport
phosphoric diester hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0