This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01800435_g1
See other MTRNAPOL GE Assays ›
Gene Symbol:
mtRNApol
Gene Name:
mitochondrial RNA polymerase
Gene Aliases:
CG4644, Dmel\CG4644, Dmel_CG4644, POLRMT, mt RNA pol, mtRNA-pol, mtRNAP
Chromosome Location:
Chr.2L: 1103254 - 1108373 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0261938
Assay ID Dm01800435_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mtRNApol
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272924.1 NP_001259853.1 3 - 4 882 82
NM_134721.3 NP_608565.1 3 - 4 882 82
GenBank mRNA BT011381.1 - 3 - 4 861 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from mitochondrial promoter
mitochondrial RNA polymerase regulatory region DNA binding
mitochondrial single subunit type RNA polymerase activity
DNA-directed RNA polymerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0