This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01799740_g1
See other NPC1B GE Assays ›
Gene Symbol:
Npc1b
Gene Name:
Niemann-Pick type C-1b
Gene Aliases:
151, 4, 7, AF2/19, CG12092, Dmel\CG12092, Dmel_CG12092, NPC1b, Q256, R-9-28, R9-28, dncp1b, l(1)19Ec, l(1)2-3, l(1)R-9-28, l151, l152, lN5, lt2-14a, npc1b
Chromosome Location:
Chr.X: 20934869 - 20939503 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0261675
Assay ID Dm01799740_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npc1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134573.3 NP_608417.2 3 - 4 2519 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s)

dorsal closure
central nervous system development
peripheral nervous system development
intestinal cholesterol absorption

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0