This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01799591_g1
See other FUR2 GE Assays ›
Gene Symbol:
Fur2
Gene Name:
Furin 2
Gene Aliases:
154521_at, CG18734, CG4235, DMH#11, Dfur2, Dfurin2, Dmel\CG18734, Dmel_CG18734, FUR2, Furin, fur2, furin
Chromosome Location:
Chr.X: 16373086 - 16381782 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004598
Assay ID Dm01799591_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fur2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001169302.2 NP_001162773.1 8 - 9 3448 70
NM_001169303.2 NP_001162774.1 10 - 11 3325 70
NM_078644.4 NP_523368.2 9 - 10 3397 70
NM_167506.3 NP_727963.1 9 - 10 3406 70
NM_167507.3 NP_727964.1 9 - 10 3506 70
NM_167508.2 NP_727965.1 9 - 10 3440 70
NM_167509.3 NP_727966.1 9 - 10 3500 70
NM_167510.3 NP_727967.1 10 - 11 3617 70
NM_206763.2 NP_996486.1 9 - 10 3384 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

protein processing
R8 cell fate specification
negative regulation of secretion
serine-type endopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0