This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01799088_m1
See other CBP GE Assays ›
Gene Symbol:
CBP
Gene Name:
sarcoplasmic calcium-binding protein
Gene Aliases:
BcDNA.GH12350, BcDNA:GH12350, CG1435, Dmel\CG1435, Dmel_CG1435, cbp, dCBP
Chromosome Location:
Chr.X: 7337352 - 7342731 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0026144
Assay ID Dm01799088_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272380.2 NP_001259309.1 2 - 3 716 59
NM_078515.4 NP_511070.1 2 - 3 716 59
NM_206638.3 NP_996361.1 2 - 3 756 59
GenBank mRNA AF104405.1 - 2 - 3 695 59
AF145675.1 - 2 - 3 698 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s)

defense response to Gram-negative bacterium

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0