This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01797007_g1
See other SEM1 GE Assays ›
Gene Symbol:
Sem1
Gene Name:
Suppressor of exocyst mutations 1
Gene Aliases:
BcDNA:RH34416, CG13779, Dmel\CG13779, Dmel_CG13779, Sem1p, dmdss1, dss1, sem1
Chromosome Location:
Chr.2L: 7063986 - 7064467 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0266666
Assay ID Dm01797007_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sem1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144298.4 NP_652555.1 1 - 2 212 76
GenBank mRNA AY070697.1 - 1 - 2 213 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
mRNA export from nucleus
proteasome-mediated ubiquitin-dependent protein catabolic process
proteasome assembly
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0