This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01796507_m1
See other PDE11 GE Assays ›
Gene Symbol:
Pde11
Gene Name:
Phosphodiesterase 11
Gene Aliases:
CG10231, CG15159, CG34341, Dmel\CG34341, Dmel_CG34341, PDE11, pde11
Chromosome Location:
Chr.2L: 18532025 - 18590342 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0085370
Assay ID Dm01796507_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103707.2 NP_001097177.1 7 - 8 2442 67
NM_001201908.2 NP_001188837.1 7 - 8 2136 67
NM_001201909.2 NP_001188838.1 7 - 8 2442 67
NM_001299115.1 NP_001286044.1 7 - 8 2309 67
NM_001299116.1 NP_001286045.1 8 - 9 1529 67
NM_136041.2 NP_609885.2 7 - 8 2195 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cAMP metabolic process
cGMP metabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0