This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01795899_m1
See other VSX2 GE Assays ›
Gene Symbol:
Vsx2
Gene Name:
Visual system homeobox 2
Gene Aliases:
CG15781, CG15782, CG15783, CG33980, Chx2, DChx2, Dmel\CG33980, Dmel_CG33980, Vsx, dChx2, dVsx2, vsx2
Chromosome Location:
Chr.X: 5528927 - 5560183 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263512
Assay ID Dm01795899_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vsx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038743.1 NP_001033832.1 3 - 4 1042 68
NM_001297968.1 NP_001284897.1 3 - 4 1042 68
GenBank mRNA BT022190.1 - 3 - 4 1042 68
BT029275.1 - 3 - 4 1042 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
optic lobe placode development
positive regulation of transcription from RNA polymerase II promoter
regulation of neural retina development
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0