Assay Details
Gene Symbol: Patronin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144219.3 | NP_001137691.1 | 9 - 10 | 1633 | 91 | |
NM_001169704.2 | NP_001163175.1 | 9 - 10 | 1633 | 91 | ||
NM_001169706.2 | NP_001163177.2 | 9 - 10 | 1633 | 91 | ||
NM_001274122.1 | NP_001261051.1 | 9 - 10 | 1633 | 91 | ||
NM_001274123.1 | NP_001261052.1 | 10 - 11 | 1786 | 91 | ||
NM_001274124.1 | NP_001261053.1 | 9 - 10 | 1681 | 91 | ||
NM_001274125.1 | NP_001261054.1 | 8 - 9 | 1528 | 91 | ||
NM_176218.3 | NP_788398.1 | 8 - 9 | 2048 | 91 | ||
NM_206154.3 | NP_995876.1 | 9 - 10 | 1633 | 91 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
