This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01794743_g1
See other PATRONIN GE Assays ›
Gene Symbol:
Patronin
Gene Name:
CG33130-PA
Gene Aliases:
BcDNA:LD17191, CG18459, CG18460, CG18462, CG30102, CG33130, CG6516, Dmel\CG33130, Dmel_CG33130, Ssp4, l(2)k07433, patronin, ssp4
Chromosome Location:
Chr.2R: 17519024 - 17535866 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263197
Assay ID Dm01794743_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Patronin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144218.3 NP_001137690.1 5 - 6 951 107
NM_001144219.3 NP_001137691.1 5 - 6 951 107
NM_001169704.2 NP_001163175.1 5 - 6 951 107
NM_001169706.2 NP_001163177.2 5 - 6 951 107
NM_001274122.1 NP_001261051.1 5 - 6 951 107
NM_001274123.1 NP_001261052.1 5 - 6 951 107
NM_001274124.1 NP_001261053.1 5 - 6 951 107
NM_001274125.1 NP_001261054.1 5 - 6 951 107
NM_001274126.1 NP_001261055.1 5 - 6 951 107
NM_176218.3 NP_788398.1 4 - 5 1366 107
NM_206153.3 NP_995875.1 5 - 6 951 107
NM_206154.3 NP_995876.1 5 - 6 951 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
cytoplasmic microtubule organization
neuron projection development
asymmetric protein localization involved in cell fate determination
establishment of mitotic spindle asymmetry
calmodulin binding
microtubule binding
spectrin binding
microtubule minus-end binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0