This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01794244_g1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID
Dm01794244_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Dscam1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001043011.1
NP_001036476.1
7 - 8
2212
75
NM_001043012.1
NP_001036477.1
7 - 8
2203
75
NM_001043013.1
NP_001036478.1
7 - 8
2203
75
NM_001043014.1
NP_001036479.1
7 - 8
2203
75
NM_001043015.1
NP_001036480.1
7 - 8
2212
75
NM_001043016.1
NP_001036481.1
7 - 8
2203
75
NM_001043017.1
NP_001036482.1
7 - 8
2212
75
NM_001043018.1
NP_001036483.1
7 - 8
2203
75
NM_001043019.1
NP_001036484.1
7 - 8
2212
75
NM_001043020.1
NP_001036485.1
7 - 8
2212
75
NM_001043021.1
NP_001036486.1
7 - 8
2209
75
NM_001043022.1
NP_001036487.1
7 - 8
2206
75
NM_001043023.1
NP_001036488.1
7 - 8
2212
75
NM_001043024.1
NP_001036489.1
7 - 8
2203
75
NM_001043025.1
NP_001036490.1
7 - 8
2212
75
NM_001043026.1
NP_001036491.1
7 - 8
2212
75
NM_001043027.1
NP_001036492.1
7 - 8
2212
75
NM_001043028.1
NP_001036493.1
7 - 8
2203
75
NM_001043029.1
NP_001036494.1
7 - 8
2203
75
NM_001043030.1
NP_001036495.1
7 - 8
2212
75
NM_001043032.1
NP_001036497.1
7 - 8
2209
75
NM_001043033.1
NP_001036498.1
7 - 8
2203
75
NM_001043034.1
NP_001036499.1
7 - 8
2212
75
NM_001043035.1
NP_001036500.1
7 - 8
2203
75
NM_001043036.1
NP_001036501.1
7 - 8
2203
75
NM_001043037.1
NP_001036502.1
7 - 8
2209
75
NM_001043038.1
NP_001036503.1
7 - 8
2203
75
NM_001043039.1
NP_001036504.1
7 - 8
2212
75
NM_001043040.1
NP_001036505.1
7 - 8
2203
75
NM_001043041.1
NP_001036506.1
7 - 8
2203
75
NM_001043044.1
NP_001036509.1
7 - 8
2203
75
NM_001043045.1
NP_001036510.1
7 - 8
2203
75
NM_001043047.1
NP_001036512.1
7 - 8
2203
75
NM_001043048.1
NP_001036513.1
7 - 8
2203
75
NM_001043049.1
NP_001036514.1
7 - 8
2203
75
NM_001043050.1
NP_001036515.1
7 - 8
2203
75
NM_001043051.1
NP_001036516.1
7 - 8
2221
75
NM_001043052.1
NP_001036517.1
7 - 8
2203
75
NM_001043053.1
NP_001036518.1
7 - 8
2203
75
NM_001043055.1
NP_001036520.1
7 - 8
2203
75
NM_001043056.1
NP_001036521.1
7 - 8
2212
75
NM_001043057.1
NP_001036522.1
7 - 8
2203
75
NM_001043058.1
NP_001036523.1
7 - 8
2203
75
NM_001043059.1
NP_001036524.1
7 - 8
2203
75
NM_001043060.1
NP_001036525.1
7 - 8
2203
75
NM_001043061.1
NP_001036526.1
7 - 8
2203
75
NM_001043062.1
NP_001036527.1
7 - 8
2200
75
NM_001043063.1
NP_001036528.1
7 - 8
2203
75
NM_001043064.1
NP_001036529.1
7 - 8
2203
75
NM_001259233.1
NP_001246162.1
7 - 8
2212
75
NM_001259234.1
NP_001246163.1
7 - 8
2203
75
NM_001259235.1
NP_001246164.1
7 - 8
2203
75
NM_001259236.1
NP_001246165.1
7 - 8
2203
75
NM_001259237.1
NP_001246166.1
7 - 8
2203
75
NM_001259238.1
NP_001246167.1
7 - 8
2203
75
NM_001259239.1
NP_001246168.1
7 - 8
2197
75
NM_001259240.1
NP_001246169.1
7 - 8
2203
75
NM_001259241.1
NP_001246170.1
7 - 8
2215
75
NM_001259242.1
NP_001246171.1
7 - 8
2212
75
NM_001259243.1
NP_001246172.1
7 - 8
2197
75
NM_001259244.1
NP_001246173.1
7 - 8
2212
75
NM_001259245.1
NP_001246174.1
7 - 8
2212
75
NM_001259246.1
NP_001246175.1
7 - 8
2209
75
NM_001259247.1
NP_001246176.1
7 - 8
2203
75
NM_001259248.1
NP_001246177.1
7 - 8
2203
75
NM_001259249.1
NP_001246178.1
7 - 8
2203
75
NM_001259250.1
NP_001246179.1
7 - 8
2212
75
NM_001259251.1
NP_001246180.1
7 - 8
2212
75
NM_001259252.1
NP_001246181.1
7 - 8
2203
75
NM_001273835.1
NP_001260764.1
7 - 8
2233
75
NM_078925.5
NP_523649.5
7 - 8
2209
75
NM_165513.1
NP_724542.1
7 - 8
2200
75
NM_165514.1
NP_724543.1
7 - 8
2200
75
NM_165515.2
NP_724544.2
7 - 8
2200
75
NM_206047.1
NP_995769.1
7 - 8
2203
75
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
G-protein coupled receptor
Drosophila melanogaster
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0