This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01794238_m1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID Dm01794238_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dscam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043011.1 NP_001036476.1 23 - 24 6547 66
NM_001043012.1 NP_001036477.1 23 - 24 6535 66
NM_001043013.1 NP_001036478.1 23 - 24 6532 66
NM_001043014.1 NP_001036479.1 23 - 24 6490 66
NM_001043015.1 NP_001036480.1 23 - 24 6499 66
NM_001043016.1 NP_001036481.1 23 - 24 6535 66
NM_001043017.1 NP_001036482.1 23 - 24 6544 66
NM_001043018.1 NP_001036483.1 23 - 24 6490 66
NM_001043019.1 NP_001036484.1 23 - 24 6544 66
NM_001043020.1 NP_001036485.1 23 - 24 6502 66
NM_001043021.1 NP_001036486.1 23 - 24 6496 66
NM_001043022.1 NP_001036487.1 23 - 24 6493 66
NM_001043023.1 NP_001036488.1 23 - 24 6502 66
NM_001043024.1 NP_001036489.1 23 - 24 6532 66
NM_001043025.1 NP_001036490.1 23 - 24 6547 66
NM_001043026.1 NP_001036491.1 23 - 24 6547 66
NM_001043027.1 NP_001036492.1 23 - 24 6544 66
NM_001043028.1 NP_001036493.1 23 - 24 6481 66
NM_001043029.1 NP_001036494.1 23 - 24 6535 66
NM_001043030.1 NP_001036495.1 23 - 24 6535 66
NM_001043032.1 NP_001036497.1 23 - 24 6496 66
NM_001043033.1 NP_001036498.1 23 - 24 6535 66
NM_001043034.1 NP_001036499.1 23 - 24 6547 66
NM_001043035.1 NP_001036500.1 23 - 24 6538 66
NM_001043036.1 NP_001036501.1 23 - 24 6490 66
NM_001043037.1 NP_001036502.1 23 - 24 6496 66
NM_001043038.1 NP_001036503.1 23 - 24 6553 66
NM_001043039.1 NP_001036504.1 23 - 24 6544 66
NM_001043040.1 NP_001036505.1 23 - 24 6532 66
NM_001043041.1 NP_001036506.1 23 - 24 6535 66
NM_001043044.1 NP_001036509.1 23 - 24 6493 66
NM_001043045.1 NP_001036510.1 23 - 24 6538 66
NM_001043047.1 NP_001036512.1 23 - 24 6493 66
NM_001043048.1 NP_001036513.1 23 - 24 6490 66
NM_001043049.1 NP_001036514.1 23 - 24 6535 66
NM_001043050.1 NP_001036515.1 23 - 24 6535 66
NM_001043051.1 NP_001036516.1 23 - 24 6508 66
NM_001043052.1 NP_001036517.1 23 - 24 6493 66
NM_001043053.1 NP_001036518.1 23 - 24 6538 66
NM_001043055.1 NP_001036520.1 23 - 24 6535 66
NM_001043056.1 NP_001036521.1 23 - 24 6499 66
NM_001043057.1 NP_001036522.1 23 - 24 6493 66
NM_001043058.1 NP_001036523.1 23 - 24 6490 66
NM_001043059.1 NP_001036524.1 23 - 24 6538 66
NM_001043060.1 NP_001036525.1 23 - 24 6538 66
NM_001043061.1 NP_001036526.1 23 - 24 6493 66
NM_001043062.1 NP_001036527.1 23 - 24 6535 66
NM_001043063.1 NP_001036528.1 23 - 24 6493 66
NM_001043064.1 NP_001036529.1 23 - 24 6490 66
NM_001259233.1 NP_001246162.1 23 - 24 6555 66
NM_001259234.1 NP_001246163.1 23 - 24 6501 66
NM_001259235.1 NP_001246164.1 23 - 24 6546 66
NM_001259236.1 NP_001246165.1 23 - 24 6546 66
NM_001259237.1 NP_001246166.1 23 - 24 6501 66
NM_001259238.1 NP_001246167.1 23 - 24 6546 66
NM_001259239.1 NP_001246168.1 23 - 24 6532 66
NM_001259240.1 NP_001246169.1 23 - 24 6541 66
NM_001259241.1 NP_001246170.1 23 - 24 6547 66
NM_001259242.1 NP_001246171.1 23 - 24 6499 66
NM_001259243.1 NP_001246172.1 23 - 24 6526 66
NM_001259244.1 NP_001246173.1 23 - 24 6550 66
NM_001259245.1 NP_001246174.1 23 - 24 6499 66
NM_001259246.1 NP_001246175.1 23 - 24 6499 66
NM_001259247.1 NP_001246176.1 23 - 24 6553 66
NM_001259248.1 NP_001246177.1 23 - 24 6538 66
NM_001259249.1 NP_001246178.1 23 - 24 6490 66
NM_001259250.1 NP_001246179.1 23 - 24 6544 66
NM_001259251.1 NP_001246180.1 23 - 24 6550 66
NM_001259252.1 NP_001246181.1 23 - 24 6490 66
NM_078925.5 NP_523649.5 23 - 24 6499 66
NM_165513.1 NP_724542.1 23 - 24 6490 66
NM_165514.1 NP_724543.1 23 - 24 6490 66
NM_165515.2 NP_724544.2 23 - 24 6490 66
NM_206047.1 NP_995769.1 23 - 24 6508 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0