This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01794237_m1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID
Dm01794237_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Dscam1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001043011.1
NP_001036476.1
22 - 23
6473
81
NM_001043012.1
NP_001036477.1
22 - 23
6461
81
NM_001043013.1
NP_001036478.1
22 - 23
6458
81
NM_001043014.1
NP_001036479.1
22 - 23
6416
81
NM_001043015.1
NP_001036480.1
22 - 23
6425
81
NM_001043016.1
NP_001036481.1
22 - 23
6461
81
NM_001043017.1
NP_001036482.1
22 - 23
6470
81
NM_001043018.1
NP_001036483.1
22 - 23
6416
81
NM_001043019.1
NP_001036484.1
22 - 23
6470
81
NM_001043020.1
NP_001036485.1
22 - 23
6428
81
NM_001043021.1
NP_001036486.1
22 - 23
6422
81
NM_001043022.1
NP_001036487.1
22 - 23
6419
81
NM_001043023.1
NP_001036488.1
22 - 23
6428
81
NM_001043024.1
NP_001036489.1
22 - 23
6458
81
NM_001043025.1
NP_001036490.1
22 - 23
6473
81
NM_001043026.1
NP_001036491.1
22 - 23
6473
81
NM_001043027.1
NP_001036492.1
22 - 23
6470
81
NM_001043028.1
NP_001036493.1
22 - 23
6407
81
NM_001043029.1
NP_001036494.1
22 - 23
6461
81
NM_001043030.1
NP_001036495.1
22 - 23
6461
81
NM_001043032.1
NP_001036497.1
22 - 23
6422
81
NM_001043033.1
NP_001036498.1
22 - 23
6461
81
NM_001043034.1
NP_001036499.1
22 - 23
6473
81
NM_001043035.1
NP_001036500.1
22 - 23
6464
81
NM_001043036.1
NP_001036501.1
22 - 23
6416
81
NM_001043037.1
NP_001036502.1
22 - 23
6422
81
NM_001043038.1
NP_001036503.1
22 - 23
6479
81
NM_001043039.1
NP_001036504.1
22 - 23
6470
81
NM_001043040.1
NP_001036505.1
22 - 23
6458
81
NM_001043041.1
NP_001036506.1
22 - 23
6461
81
NM_001043044.1
NP_001036509.1
22 - 23
6419
81
NM_001043045.1
NP_001036510.1
22 - 23
6464
81
NM_001043047.1
NP_001036512.1
22 - 23
6419
81
NM_001043048.1
NP_001036513.1
22 - 23
6416
81
NM_001043049.1
NP_001036514.1
22 - 23
6461
81
NM_001043050.1
NP_001036515.1
22 - 23
6461
81
NM_001043051.1
NP_001036516.1
22 - 23
6434
81
NM_001043052.1
NP_001036517.1
22 - 23
6419
81
NM_001043053.1
NP_001036518.1
22 - 23
6464
81
NM_001043055.1
NP_001036520.1
22 - 23
6461
81
NM_001043056.1
NP_001036521.1
22 - 23
6425
81
NM_001043057.1
NP_001036522.1
22 - 23
6419
81
NM_001043058.1
NP_001036523.1
22 - 23
6416
81
NM_001043059.1
NP_001036524.1
22 - 23
6464
81
NM_001043060.1
NP_001036525.1
22 - 23
6464
81
NM_001043061.1
NP_001036526.1
22 - 23
6419
81
NM_001043062.1
NP_001036527.1
22 - 23
6461
81
NM_001043063.1
NP_001036528.1
22 - 23
6419
81
NM_001043064.1
NP_001036529.1
22 - 23
6416
81
NM_001259233.1
NP_001246162.1
22 - 23
6481
81
NM_001259234.1
NP_001246163.1
22 - 23
6427
81
NM_001259235.1
NP_001246164.1
22 - 23
6472
81
NM_001259236.1
NP_001246165.1
22 - 23
6472
81
NM_001259237.1
NP_001246166.1
22 - 23
6427
81
NM_001259238.1
NP_001246167.1
22 - 23
6472
81
NM_001259239.1
NP_001246168.1
22 - 23
6458
81
NM_001259240.1
NP_001246169.1
22 - 23
6467
81
NM_001259241.1
NP_001246170.1
22 - 23
6473
81
NM_001259242.1
NP_001246171.1
22 - 23
6425
81
NM_001259243.1
NP_001246172.1
22 - 23
6452
81
NM_001259244.1
NP_001246173.1
22 - 23
6476
81
NM_001259245.1
NP_001246174.1
22 - 23
6425
81
NM_001259246.1
NP_001246175.1
22 - 23
6425
81
NM_001259247.1
NP_001246176.1
22 - 23
6479
81
NM_001259248.1
NP_001246177.1
22 - 23
6464
81
NM_001259249.1
NP_001246178.1
22 - 23
6416
81
NM_001259250.1
NP_001246179.1
22 - 23
6470
81
NM_001259251.1
NP_001246180.1
22 - 23
6476
81
NM_001259252.1
NP_001246181.1
22 - 23
6416
81
NM_078925.5
NP_523649.5
22 - 23
6425
81
NM_165513.1
NP_724542.1
22 - 23
6416
81
NM_165514.1
NP_724543.1
22 - 23
6416
81
NM_165515.2
NP_724544.2
22 - 23
6416
81
NM_206047.1
NP_995769.1
22 - 23
6434
81
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
G-protein coupled receptor
Drosophila melanogaster
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0