This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01794235_g1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID Dm01794235_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dscam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043011.1 NP_001036476.1 20 - 21 6140 59
NM_001043012.1 NP_001036477.1 20 - 21 6128 59
NM_001043013.1 NP_001036478.1 20 - 21 6125 59
NM_001043014.1 NP_001036479.1 20 - 21 6083 59
NM_001043015.1 NP_001036480.1 20 - 21 6092 59
NM_001043016.1 NP_001036481.1 20 - 21 6128 59
NM_001043017.1 NP_001036482.1 20 - 21 6137 59
NM_001043018.1 NP_001036483.1 20 - 21 6083 59
NM_001043019.1 NP_001036484.1 20 - 21 6137 59
NM_001043020.1 NP_001036485.1 20 - 21 6095 59
NM_001043021.1 NP_001036486.1 20 - 21 6089 59
NM_001043022.1 NP_001036487.1 20 - 21 6086 59
NM_001043023.1 NP_001036488.1 20 - 21 6095 59
NM_001043024.1 NP_001036489.1 20 - 21 6125 59
NM_001043025.1 NP_001036490.1 20 - 21 6140 59
NM_001043026.1 NP_001036491.1 20 - 21 6140 59
NM_001043027.1 NP_001036492.1 20 - 21 6137 59
NM_001043028.1 NP_001036493.1 20 - 21 6074 59
NM_001043029.1 NP_001036494.1 20 - 21 6128 59
NM_001043030.1 NP_001036495.1 20 - 21 6128 59
NM_001043032.1 NP_001036497.1 20 - 21 6089 59
NM_001043033.1 NP_001036498.1 20 - 21 6128 59
NM_001043034.1 NP_001036499.1 20 - 21 6140 59
NM_001043035.1 NP_001036500.1 20 - 21 6131 59
NM_001043036.1 NP_001036501.1 20 - 21 6083 59
NM_001043037.1 NP_001036502.1 20 - 21 6089 59
NM_001043038.1 NP_001036503.1 20 - 21 6146 59
NM_001043039.1 NP_001036504.1 20 - 21 6137 59
NM_001043040.1 NP_001036505.1 20 - 21 6125 59
NM_001043041.1 NP_001036506.1 20 - 21 6128 59
NM_001043044.1 NP_001036509.1 20 - 21 6086 59
NM_001043045.1 NP_001036510.1 20 - 21 6131 59
NM_001043047.1 NP_001036512.1 20 - 21 6086 59
NM_001043048.1 NP_001036513.1 20 - 21 6083 59
NM_001043049.1 NP_001036514.1 20 - 21 6128 59
NM_001043050.1 NP_001036515.1 20 - 21 6128 59
NM_001043051.1 NP_001036516.1 20 - 21 6101 59
NM_001043052.1 NP_001036517.1 20 - 21 6086 59
NM_001043053.1 NP_001036518.1 20 - 21 6131 59
NM_001043055.1 NP_001036520.1 20 - 21 6128 59
NM_001043056.1 NP_001036521.1 20 - 21 6092 59
NM_001043057.1 NP_001036522.1 20 - 21 6086 59
NM_001043058.1 NP_001036523.1 20 - 21 6083 59
NM_001043059.1 NP_001036524.1 20 - 21 6131 59
NM_001043060.1 NP_001036525.1 20 - 21 6131 59
NM_001043061.1 NP_001036526.1 20 - 21 6086 59
NM_001043062.1 NP_001036527.1 20 - 21 6128 59
NM_001043063.1 NP_001036528.1 20 - 21 6086 59
NM_001043064.1 NP_001036529.1 20 - 21 6083 59
NM_001259233.1 NP_001246162.1 20 - 21 6148 59
NM_001259234.1 NP_001246163.1 20 - 21 6094 59
NM_001259235.1 NP_001246164.1 20 - 21 6139 59
NM_001259236.1 NP_001246165.1 20 - 21 6139 59
NM_001259237.1 NP_001246166.1 20 - 21 6094 59
NM_001259238.1 NP_001246167.1 20 - 21 6139 59
NM_001259239.1 NP_001246168.1 20 - 21 6125 59
NM_001259240.1 NP_001246169.1 20 - 21 6134 59
NM_001259241.1 NP_001246170.1 20 - 21 6140 59
NM_001259242.1 NP_001246171.1 20 - 21 6092 59
NM_001259243.1 NP_001246172.1 20 - 21 6119 59
NM_001259244.1 NP_001246173.1 20 - 21 6143 59
NM_001259245.1 NP_001246174.1 20 - 21 6092 59
NM_001259246.1 NP_001246175.1 20 - 21 6092 59
NM_001259247.1 NP_001246176.1 20 - 21 6146 59
NM_001259248.1 NP_001246177.1 20 - 21 6131 59
NM_001259249.1 NP_001246178.1 20 - 21 6083 59
NM_001259250.1 NP_001246179.1 20 - 21 6137 59
NM_001259251.1 NP_001246180.1 20 - 21 6143 59
NM_001259252.1 NP_001246181.1 20 - 21 6083 59
NM_001273835.1 NP_001260764.1 19 - 20 5966 59
NM_078925.5 NP_523649.5 20 - 21 6092 59
NM_165513.1 NP_724542.1 20 - 21 6083 59
NM_165514.1 NP_724543.1 20 - 21 6083 59
NM_165515.2 NP_724544.2 20 - 21 6083 59
NM_206047.1 NP_995769.1 20 - 21 6101 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0