This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01794233_g1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID Dm01794233_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dscam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043011.1 NP_001036476.1 19 - 20 5825 70
NM_001043012.1 NP_001036477.1 19 - 20 5813 70
NM_001043013.1 NP_001036478.1 19 - 20 5810 70
NM_001043014.1 NP_001036479.1 19 - 20 5768 70
NM_001043015.1 NP_001036480.1 19 - 20 5777 70
NM_001043016.1 NP_001036481.1 19 - 20 5813 70
NM_001043017.1 NP_001036482.1 19 - 20 5822 70
NM_001043018.1 NP_001036483.1 19 - 20 5768 70
NM_001043019.1 NP_001036484.1 19 - 20 5822 70
NM_001043020.1 NP_001036485.1 19 - 20 5780 70
NM_001043021.1 NP_001036486.1 19 - 20 5774 70
NM_001043022.1 NP_001036487.1 19 - 20 5771 70
NM_001043023.1 NP_001036488.1 19 - 20 5780 70
NM_001043024.1 NP_001036489.1 19 - 20 5810 70
NM_001043025.1 NP_001036490.1 19 - 20 5825 70
NM_001043026.1 NP_001036491.1 19 - 20 5825 70
NM_001043027.1 NP_001036492.1 19 - 20 5822 70
NM_001043028.1 NP_001036493.1 19 - 20 5759 70
NM_001043029.1 NP_001036494.1 19 - 20 5813 70
NM_001043030.1 NP_001036495.1 19 - 20 5813 70
NM_001043032.1 NP_001036497.1 19 - 20 5774 70
NM_001043033.1 NP_001036498.1 19 - 20 5813 70
NM_001043034.1 NP_001036499.1 19 - 20 5825 70
NM_001043035.1 NP_001036500.1 19 - 20 5816 70
NM_001043036.1 NP_001036501.1 19 - 20 5768 70
NM_001043037.1 NP_001036502.1 19 - 20 5774 70
NM_001043038.1 NP_001036503.1 19 - 20 5831 70
NM_001043039.1 NP_001036504.1 19 - 20 5822 70
NM_001043040.1 NP_001036505.1 19 - 20 5810 70
NM_001043041.1 NP_001036506.1 19 - 20 5813 70
NM_001043044.1 NP_001036509.1 19 - 20 5771 70
NM_001043045.1 NP_001036510.1 19 - 20 5816 70
NM_001043047.1 NP_001036512.1 19 - 20 5771 70
NM_001043048.1 NP_001036513.1 19 - 20 5768 70
NM_001043049.1 NP_001036514.1 19 - 20 5813 70
NM_001043050.1 NP_001036515.1 19 - 20 5813 70
NM_001043051.1 NP_001036516.1 19 - 20 5786 70
NM_001043052.1 NP_001036517.1 19 - 20 5771 70
NM_001043053.1 NP_001036518.1 19 - 20 5816 70
NM_001043055.1 NP_001036520.1 19 - 20 5813 70
NM_001043056.1 NP_001036521.1 19 - 20 5777 70
NM_001043057.1 NP_001036522.1 19 - 20 5771 70
NM_001043058.1 NP_001036523.1 19 - 20 5768 70
NM_001043059.1 NP_001036524.1 19 - 20 5816 70
NM_001043060.1 NP_001036525.1 19 - 20 5816 70
NM_001043061.1 NP_001036526.1 19 - 20 5771 70
NM_001043062.1 NP_001036527.1 19 - 20 5813 70
NM_001043063.1 NP_001036528.1 19 - 20 5771 70
NM_001043064.1 NP_001036529.1 19 - 20 5768 70
NM_001259233.1 NP_001246162.1 19 - 20 5833 70
NM_001259234.1 NP_001246163.1 19 - 20 5779 70
NM_001259235.1 NP_001246164.1 19 - 20 5824 70
NM_001259236.1 NP_001246165.1 19 - 20 5824 70
NM_001259237.1 NP_001246166.1 19 - 20 5779 70
NM_001259238.1 NP_001246167.1 19 - 20 5824 70
NM_001259239.1 NP_001246168.1 19 - 20 5810 70
NM_001259240.1 NP_001246169.1 19 - 20 5819 70
NM_001259241.1 NP_001246170.1 19 - 20 5825 70
NM_001259242.1 NP_001246171.1 19 - 20 5777 70
NM_001259243.1 NP_001246172.1 19 - 20 5804 70
NM_001259244.1 NP_001246173.1 19 - 20 5828 70
NM_001259245.1 NP_001246174.1 19 - 20 5777 70
NM_001259246.1 NP_001246175.1 19 - 20 5777 70
NM_001259247.1 NP_001246176.1 19 - 20 5831 70
NM_001259248.1 NP_001246177.1 19 - 20 5816 70
NM_001259249.1 NP_001246178.1 19 - 20 5768 70
NM_001259250.1 NP_001246179.1 19 - 20 5822 70
NM_001259251.1 NP_001246180.1 19 - 20 5828 70
NM_001259252.1 NP_001246181.1 19 - 20 5768 70
NM_078925.5 NP_523649.5 19 - 20 5777 70
NM_165513.1 NP_724542.1 19 - 20 5768 70
NM_165514.1 NP_724543.1 19 - 20 5768 70
NM_165515.2 NP_724544.2 19 - 20 5768 70
NM_206047.1 NP_995769.1 19 - 20 5786 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0