This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01794229_g1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID
Dm01794229_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Dscam1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001043011.1
NP_001036476.1
15 - 16
5147
73
NM_001043012.1
NP_001036477.1
15 - 16
5135
73
NM_001043013.1
NP_001036478.1
15 - 16
5132
73
NM_001043014.1
NP_001036479.1
15 - 16
5135
73
NM_001043015.1
NP_001036480.1
15 - 16
5144
73
NM_001043016.1
NP_001036481.1
15 - 16
5135
73
NM_001043017.1
NP_001036482.1
15 - 16
5144
73
NM_001043018.1
NP_001036483.1
15 - 16
5135
73
NM_001043019.1
NP_001036484.1
15 - 16
5144
73
NM_001043020.1
NP_001036485.1
15 - 16
5147
73
NM_001043021.1
NP_001036486.1
15 - 16
5141
73
NM_001043022.1
NP_001036487.1
15 - 16
5138
73
NM_001043023.1
NP_001036488.1
15 - 16
5147
73
NM_001043024.1
NP_001036489.1
15 - 16
5132
73
NM_001043025.1
NP_001036490.1
15 - 16
5147
73
NM_001043026.1
NP_001036491.1
15 - 16
5147
73
NM_001043027.1
NP_001036492.1
15 - 16
5144
73
NM_001043028.1
NP_001036493.1
15 - 16
5126
73
NM_001043029.1
NP_001036494.1
15 - 16
5135
73
NM_001043030.1
NP_001036495.1
15 - 16
5135
73
NM_001043032.1
NP_001036497.1
15 - 16
5141
73
NM_001043033.1
NP_001036498.1
15 - 16
5135
73
NM_001043034.1
NP_001036499.1
15 - 16
5147
73
NM_001043035.1
NP_001036500.1
15 - 16
5138
73
NM_001043036.1
NP_001036501.1
15 - 16
5135
73
NM_001043037.1
NP_001036502.1
15 - 16
5141
73
NM_001043038.1
NP_001036503.1
15 - 16
5153
73
NM_001043039.1
NP_001036504.1
15 - 16
5144
73
NM_001043040.1
NP_001036505.1
15 - 16
5132
73
NM_001043041.1
NP_001036506.1
15 - 16
5135
73
NM_001043044.1
NP_001036509.1
15 - 16
5138
73
NM_001043045.1
NP_001036510.1
15 - 16
5138
73
NM_001043047.1
NP_001036512.1
15 - 16
5138
73
NM_001043048.1
NP_001036513.1
15 - 16
5135
73
NM_001043049.1
NP_001036514.1
15 - 16
5135
73
NM_001043050.1
NP_001036515.1
15 - 16
5135
73
NM_001043051.1
NP_001036516.1
15 - 16
5153
73
NM_001043052.1
NP_001036517.1
15 - 16
5138
73
NM_001043053.1
NP_001036518.1
15 - 16
5138
73
NM_001043055.1
NP_001036520.1
15 - 16
5135
73
NM_001043056.1
NP_001036521.1
15 - 16
5144
73
NM_001043057.1
NP_001036522.1
15 - 16
5138
73
NM_001043058.1
NP_001036523.1
15 - 16
5135
73
NM_001043059.1
NP_001036524.1
15 - 16
5138
73
NM_001043060.1
NP_001036525.1
15 - 16
5138
73
NM_001043061.1
NP_001036526.1
15 - 16
5138
73
NM_001043062.1
NP_001036527.1
15 - 16
5135
73
NM_001043063.1
NP_001036528.1
15 - 16
5138
73
NM_001043064.1
NP_001036529.1
15 - 16
5135
73
NM_001259233.1
NP_001246162.1
15 - 16
5155
73
NM_001259234.1
NP_001246163.1
15 - 16
5146
73
NM_001259235.1
NP_001246164.1
15 - 16
5146
73
NM_001259236.1
NP_001246165.1
15 - 16
5146
73
NM_001259237.1
NP_001246166.1
15 - 16
5146
73
NM_001259238.1
NP_001246167.1
15 - 16
5146
73
NM_001259239.1
NP_001246168.1
15 - 16
5132
73
NM_001259240.1
NP_001246169.1
15 - 16
5141
73
NM_001259241.1
NP_001246170.1
15 - 16
5147
73
NM_001259242.1
NP_001246171.1
15 - 16
5144
73
NM_001259243.1
NP_001246172.1
15 - 16
5126
73
NM_001259244.1
NP_001246173.1
15 - 16
5150
73
NM_001259245.1
NP_001246174.1
15 - 16
5144
73
NM_001259246.1
NP_001246175.1
15 - 16
5144
73
NM_001259247.1
NP_001246176.1
15 - 16
5153
73
NM_001259248.1
NP_001246177.1
15 - 16
5138
73
NM_001259249.1
NP_001246178.1
15 - 16
5135
73
NM_001259250.1
NP_001246179.1
15 - 16
5144
73
NM_001259251.1
NP_001246180.1
15 - 16
5150
73
NM_001259252.1
NP_001246181.1
15 - 16
5135
73
NM_001273835.1
NP_001260764.1
15 - 16
5165
73
NM_078925.5
NP_523649.5
15 - 16
5144
73
NM_165513.1
NP_724542.1
15 - 16
5135
73
NM_165514.1
NP_724543.1
15 - 16
5135
73
NM_165515.2
NP_724544.2
15 - 16
5135
73
NM_206047.1
NP_995769.1
15 - 16
5153
73
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
G-protein coupled receptor
Drosophila melanogaster
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0