This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01794226_g1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID Dm01794226_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dscam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043011.1 NP_001036476.1 12 - 13 4175 83
NM_001043012.1 NP_001036477.1 12 - 13 4163 83
NM_001043013.1 NP_001036478.1 12 - 13 4160 83
NM_001043014.1 NP_001036479.1 12 - 13 4163 83
NM_001043015.1 NP_001036480.1 12 - 13 4172 83
NM_001043016.1 NP_001036481.1 12 - 13 4163 83
NM_001043017.1 NP_001036482.1 12 - 13 4172 83
NM_001043018.1 NP_001036483.1 12 - 13 4163 83
NM_001043019.1 NP_001036484.1 12 - 13 4172 83
NM_001043020.1 NP_001036485.1 12 - 13 4175 83
NM_001043021.1 NP_001036486.1 12 - 13 4169 83
NM_001043022.1 NP_001036487.1 12 - 13 4166 83
NM_001043023.1 NP_001036488.1 12 - 13 4175 83
NM_001043024.1 NP_001036489.1 12 - 13 4160 83
NM_001043025.1 NP_001036490.1 12 - 13 4175 83
NM_001043026.1 NP_001036491.1 12 - 13 4175 83
NM_001043027.1 NP_001036492.1 12 - 13 4172 83
NM_001043028.1 NP_001036493.1 12 - 13 4154 83
NM_001043029.1 NP_001036494.1 12 - 13 4163 83
NM_001043030.1 NP_001036495.1 12 - 13 4163 83
NM_001043032.1 NP_001036497.1 12 - 13 4169 83
NM_001043033.1 NP_001036498.1 12 - 13 4163 83
NM_001043034.1 NP_001036499.1 12 - 13 4175 83
NM_001043035.1 NP_001036500.1 12 - 13 4166 83
NM_001043036.1 NP_001036501.1 12 - 13 4163 83
NM_001043037.1 NP_001036502.1 12 - 13 4169 83
NM_001043038.1 NP_001036503.1 12 - 13 4181 83
NM_001043039.1 NP_001036504.1 12 - 13 4172 83
NM_001043040.1 NP_001036505.1 12 - 13 4160 83
NM_001043041.1 NP_001036506.1 12 - 13 4163 83
NM_001043044.1 NP_001036509.1 12 - 13 4166 83
NM_001043045.1 NP_001036510.1 12 - 13 4166 83
NM_001043047.1 NP_001036512.1 12 - 13 4166 83
NM_001043048.1 NP_001036513.1 12 - 13 4163 83
NM_001043049.1 NP_001036514.1 12 - 13 4163 83
NM_001043050.1 NP_001036515.1 12 - 13 4163 83
NM_001043051.1 NP_001036516.1 12 - 13 4181 83
NM_001043052.1 NP_001036517.1 12 - 13 4166 83
NM_001043053.1 NP_001036518.1 12 - 13 4166 83
NM_001043055.1 NP_001036520.1 12 - 13 4163 83
NM_001043056.1 NP_001036521.1 12 - 13 4172 83
NM_001043057.1 NP_001036522.1 12 - 13 4166 83
NM_001043058.1 NP_001036523.1 12 - 13 4163 83
NM_001043059.1 NP_001036524.1 12 - 13 4166 83
NM_001043060.1 NP_001036525.1 12 - 13 4166 83
NM_001043061.1 NP_001036526.1 12 - 13 4166 83
NM_001043062.1 NP_001036527.1 12 - 13 4163 83
NM_001043063.1 NP_001036528.1 12 - 13 4166 83
NM_001043064.1 NP_001036529.1 12 - 13 4163 83
NM_001259233.1 NP_001246162.1 12 - 13 4183 83
NM_001259234.1 NP_001246163.1 12 - 13 4174 83
NM_001259235.1 NP_001246164.1 12 - 13 4174 83
NM_001259236.1 NP_001246165.1 12 - 13 4174 83
NM_001259237.1 NP_001246166.1 12 - 13 4174 83
NM_001259238.1 NP_001246167.1 12 - 13 4174 83
NM_001259239.1 NP_001246168.1 12 - 13 4160 83
NM_001259240.1 NP_001246169.1 12 - 13 4169 83
NM_001259241.1 NP_001246170.1 12 - 13 4175 83
NM_001259242.1 NP_001246171.1 12 - 13 4172 83
NM_001259243.1 NP_001246172.1 12 - 13 4154 83
NM_001259244.1 NP_001246173.1 12 - 13 4178 83
NM_001259245.1 NP_001246174.1 12 - 13 4172 83
NM_001259246.1 NP_001246175.1 12 - 13 4172 83
NM_001259247.1 NP_001246176.1 12 - 13 4181 83
NM_001259248.1 NP_001246177.1 12 - 13 4166 83
NM_001259249.1 NP_001246178.1 12 - 13 4163 83
NM_001259250.1 NP_001246179.1 12 - 13 4172 83
NM_001259251.1 NP_001246180.1 12 - 13 4178 83
NM_001259252.1 NP_001246181.1 12 - 13 4163 83
NM_001273835.1 NP_001260764.1 12 - 13 4193 83
NM_078925.5 NP_523649.5 12 - 13 4172 83
NM_165513.1 NP_724542.1 12 - 13 4163 83
NM_165514.1 NP_724543.1 12 - 13 4163 83
NM_165515.2 NP_724544.2 12 - 13 4163 83
NM_206047.1 NP_995769.1 12 - 13 4181 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0