This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01794225_g1
See other DSCAM1 GE Assays ›
Gene Symbol:
Dscam1
Gene Name:
Down syndrome cell adhesion molecule 1
Gene Aliases:
43Bc, CG17800, CT39257, DSCAM, DScam, DmDscam, Dm_2R:13579, Dm_2R:13612, Dmel\CG17800, Dmel_CG17800, DsCam, Dscam, Dscam-hv, Dsm, Neu1, dScam, dscam, dscam1, l(2)05518, l(2)43Bc, p270
Chromosome Location:
Chr.2R: 7317924 - 7381899 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033159
Assay ID
Dm01794225_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Dscam1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001043011.1
NP_001036476.1
11 - 12
4019
94
NM_001043012.1
NP_001036477.1
11 - 12
4007
94
NM_001043013.1
NP_001036478.1
11 - 12
4004
94
NM_001043014.1
NP_001036479.1
11 - 12
4007
94
NM_001043015.1
NP_001036480.1
11 - 12
4016
94
NM_001043016.1
NP_001036481.1
11 - 12
4007
94
NM_001043017.1
NP_001036482.1
11 - 12
4016
94
NM_001043018.1
NP_001036483.1
11 - 12
4007
94
NM_001043019.1
NP_001036484.1
11 - 12
4016
94
NM_001043020.1
NP_001036485.1
11 - 12
4019
94
NM_001043021.1
NP_001036486.1
11 - 12
4013
94
NM_001043022.1
NP_001036487.1
11 - 12
4010
94
NM_001043023.1
NP_001036488.1
11 - 12
4019
94
NM_001043024.1
NP_001036489.1
11 - 12
4004
94
NM_001043025.1
NP_001036490.1
11 - 12
4019
94
NM_001043026.1
NP_001036491.1
11 - 12
4019
94
NM_001043027.1
NP_001036492.1
11 - 12
4016
94
NM_001043028.1
NP_001036493.1
11 - 12
3998
94
NM_001043029.1
NP_001036494.1
11 - 12
4007
94
NM_001043030.1
NP_001036495.1
11 - 12
4007
94
NM_001043032.1
NP_001036497.1
11 - 12
4013
94
NM_001043033.1
NP_001036498.1
11 - 12
4007
94
NM_001043034.1
NP_001036499.1
11 - 12
4019
94
NM_001043035.1
NP_001036500.1
11 - 12
4010
94
NM_001043036.1
NP_001036501.1
11 - 12
4007
94
NM_001043037.1
NP_001036502.1
11 - 12
4013
94
NM_001043038.1
NP_001036503.1
11 - 12
4025
94
NM_001043039.1
NP_001036504.1
11 - 12
4016
94
NM_001043040.1
NP_001036505.1
11 - 12
4004
94
NM_001043041.1
NP_001036506.1
11 - 12
4007
94
NM_001043044.1
NP_001036509.1
11 - 12
4010
94
NM_001043045.1
NP_001036510.1
11 - 12
4010
94
NM_001043047.1
NP_001036512.1
11 - 12
4010
94
NM_001043048.1
NP_001036513.1
11 - 12
4007
94
NM_001043049.1
NP_001036514.1
11 - 12
4007
94
NM_001043050.1
NP_001036515.1
11 - 12
4007
94
NM_001043051.1
NP_001036516.1
11 - 12
4025
94
NM_001043052.1
NP_001036517.1
11 - 12
4010
94
NM_001043053.1
NP_001036518.1
11 - 12
4010
94
NM_001043055.1
NP_001036520.1
11 - 12
4007
94
NM_001043056.1
NP_001036521.1
11 - 12
4016
94
NM_001043057.1
NP_001036522.1
11 - 12
4010
94
NM_001043058.1
NP_001036523.1
11 - 12
4007
94
NM_001043059.1
NP_001036524.1
11 - 12
4010
94
NM_001043060.1
NP_001036525.1
11 - 12
4010
94
NM_001043061.1
NP_001036526.1
11 - 12
4010
94
NM_001043062.1
NP_001036527.1
11 - 12
4007
94
NM_001043063.1
NP_001036528.1
11 - 12
4010
94
NM_001043064.1
NP_001036529.1
11 - 12
4007
94
NM_001259233.1
NP_001246162.1
11 - 12
4027
94
NM_001259234.1
NP_001246163.1
11 - 12
4018
94
NM_001259235.1
NP_001246164.1
11 - 12
4018
94
NM_001259236.1
NP_001246165.1
11 - 12
4018
94
NM_001259237.1
NP_001246166.1
11 - 12
4018
94
NM_001259238.1
NP_001246167.1
11 - 12
4018
94
NM_001259239.1
NP_001246168.1
11 - 12
4004
94
NM_001259240.1
NP_001246169.1
11 - 12
4013
94
NM_001259241.1
NP_001246170.1
11 - 12
4019
94
NM_001259242.1
NP_001246171.1
11 - 12
4016
94
NM_001259243.1
NP_001246172.1
11 - 12
3998
94
NM_001259244.1
NP_001246173.1
11 - 12
4022
94
NM_001259245.1
NP_001246174.1
11 - 12
4016
94
NM_001259246.1
NP_001246175.1
11 - 12
4016
94
NM_001259247.1
NP_001246176.1
11 - 12
4025
94
NM_001259248.1
NP_001246177.1
11 - 12
4010
94
NM_001259249.1
NP_001246178.1
11 - 12
4007
94
NM_001259250.1
NP_001246179.1
11 - 12
4016
94
NM_001259251.1
NP_001246180.1
11 - 12
4022
94
NM_001259252.1
NP_001246181.1
11 - 12
4007
94
NM_001273835.1
NP_001260764.1
11 - 12
4037
94
NM_078925.5
NP_523649.5
11 - 12
4016
94
NM_165513.1
NP_724542.1
11 - 12
4007
94
NM_165514.1
NP_724543.1
11 - 12
4007
94
NM_165515.2
NP_724544.2
11 - 12
4007
94
NM_206047.1
NP_995769.1
11 - 12
4025
94
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
G-protein coupled receptor
Drosophila melanogaster
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
mushroom body development
central nervous system morphogenesis
positive regulation of cell migration
detection of molecule of bacterial origin
neuron development
regulation of dendrite morphogenesis
axon extension involved in axon guidance
regulation of axonogenesis
detection of mechanical stimulus involved in sensory perception of touch
dendrite self-avoidance
antigen binding
axon guidance receptor activity
identical protein binding
protein homodimerization activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0