This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01793712_m1
See other FTZ-F1 GE Assays ›
Gene Symbol:
ftz-f1
Gene Name:
ftz transcription factor 1
Gene Aliases:
BFTZ-F1, CG13378, CG4059, DEP1, DmFTZ-F1, Dmel\CG4059, Dmel_CG4059, FIP1, FTZ-F1, FTZ-F1A, FTZ-F1alpha, FTZ-Fl, FTZF1, FTZf1alpha, Ftz-F1, Ftz-f1, Ftzf1, NR5A3, alphaFTZ-F1, alphaFTZF1, alphaftz-f1, bFTZ-F1, beta FTZ-F1, beta Ftz-f1, beta-FTZ-F1, beta-Ftz-F1, betaFTZ-F1, betaFTZF1, betaFtz-F1, betaFtz-f1, betaftz-F1, betaftz-f1, betaftzf1, dFTZ-F1alpha, dFtz-F1alpha, dmFTZ-f1, ftz-F1, ftz-f1alpha, ftzf1, l(3)03649
Chromosome Location:
Chr.3L: 18750607 - 18801309 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0001078
Assay ID Dm01793712_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ftz-f1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259895.2 NP_001246824.1 1 - 2 2415 79
NM_001316437.1 NP_001303366.1 1 - 2 2415 79
NM_079419.3 NP_524143.2 3 - 4 3054 79
NM_168775.2 NP_730359.1 1 - 2 1627 79
GenBank mRNA BT058016.1 - 3 - 4 2640 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

instar larval or pupal development
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
imaginal disc-derived leg morphogenesis
metamorphosis
cell death
response to hormone
hormone-mediated signaling pathway
tissue development
mushroom body development
neuron remodeling
pupariation
pupation
response to ecdysone
juvenile hormone mediated signaling pathway
regulation of development, heterochronic
positive regulation of transcription from RNA polymerase II promoter
dendrite morphogenesis
lipid homeostasis
transcription regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
transcription factor binding
zinc ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0