This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01792295_g1
See other EST-Q GE Assays ›
Gene Symbol:
Est-Q
Gene Name:
Esterase Q
Gene Aliases:
CG7529, Dmel\CG7529, Dmel_CG7529
Chromosome Location:
Chr.3L: 21501421 - 21503959 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0037090
Assay ID Dm01792295_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Est-Q
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_141064.3 NP_649321.1 4 - 5 1283 97
GenBank mRNA AY051599.1 - 4 - 5 1248 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

hydrolase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0