This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01791319_g1
See other NIMA GE Assays ›
Gene Symbol:
NimA
Gene Name:
Nimrod A
Gene Aliases:
BG:DS00180.5, CG31765, CG31841, CG42282, CG8855, CT25428, DS00180.5, Dmel\CG42282, Dmel_CG42282, j, nimA
Chromosome Location:
Chr.2L: 13957539 - 13963381 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0261514
Assay ID Dm01791319_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NimA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259097.1 NP_001246026.1 3 - 4 910 79
NM_001298989.1 NP_001285918.1 3 - 4 664 79
NM_135847.6 NP_609691.6 3 - 4 910 79
GenBank mRNA BT100252.1 - 3 - 4 910 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

membrane-bound signaling molecule

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0