This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cp04183079_m1
See other SVP GE Assays ›
Gene Symbol:
Svp
Gene Name:
seminal vesicle polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.Un|NT_176374: 11125107 - 11279709 on Build Cavpor3.0
Species:
Guinea pig
Species Specific ID (Flybase ID):
-
Assay ID Cp04183079_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Svp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172730.1 NP_001166201.1 4 - 5 739 104
GenBank mRNA AF001982.1 - 2 - 3 409 104
AF002657.1 - 4 - 5 739 104
AF002658.1 - 2 - 3 521 104
M33295.1 - 1 - 2 58 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Cavia porcellus Probe spans exons

Panther Classification:

Molecular Function -

protease inhibitor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0