This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cp03755474_m1
See other CD46 GE Assays ›
Gene Symbol:
Cd46
Gene Name:
CD46 molecule
Gene Aliases:
MCP
Chromosome Location:
Chr.Un|NT_176407: 15332005 - 15365185 on Build Cavpor3.0
Species:
Guinea pig
Species Specific ID (Flybase ID):
-
Assay ID Cp03755474_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd46
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172928.2 NP_001166399.2 3 - 4 404 88
XM_005005806.3 XP_005005863.1 3 - 4 506 88
XM_013157800.2 XP_013013254.1 3 - 4 506 88
XM_013157801.2 XP_013013255.1 3 - 4 506 88
XM_013157802.2 XP_013013256.1 3 - 4 506 88
GenBank mRNA D84130.1 - 3 - 4 342 88
D84131.1 - 3 - 4 404 88
D84132.1 - 3 - 4 479 88
D84133.1 - 3 - 4 492 88
D88358.1 - 3 - 4 404 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Cavia porcellus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0