Assay Details
Gene Symbol: CXHXorf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008989459.2 | XP_008987707.1 | 4 - 5 | 672 | 70 | |
XM_017968721.1 | XP_017824210.1 | 3 - 4 | 584 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
