Assay Details
Gene Symbol: LOC100399524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008984529.2 | XP_008982777.1 | 4 - 5 | 619 | 76 | |
XM_008984530.1 | XP_008982778.1 | 4 - 5 | 619 | 76 | ||
XM_008984531.2 | XP_008982779.1 | 4 - 5 | 606 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
