This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Cj06107210_m1
See other ASPSCR1 GE Assays ›
Gene Symbol:
ASPSCR1
Gene Name:
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases:
-
Chromosome Location:
Chr.5: 131112702 - 131160441 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID
Cj06107210_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ASPSCR1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_008997928.2
XP_008996176.1
10 - 11
1222
66
XM_008997930.2
XP_008996178.1
10 - 11
1218
66
XM_008997931.2
XP_008996179.1
10 - 11
1221
66
XM_008997932.2
XP_008996180.1
10 - 11
1222
66
XM_008997933.2
XP_008996181.1
10 - 11
1213
66
XM_008997934.2
XP_008996182.1
10 - 11
1216
66
XM_008997935.2
XP_008996183.1
10 - 11
1227
66
XM_008997936.2
XP_008996184.1
10 - 11
1217
66
XM_008997937.2
XP_008996185.1
10 - 11
1225
66
XM_008997938.2
XP_008996186.1
10 - 11
1222
66
XM_008997939.2
XP_008996187.1
10 - 11
1221
66
XM_017972936.1
XP_017828425.1
7 - 8
1150
66
XM_017972937.1
XP_017828426.1
9 - 10
1069
66
XM_017972938.1
XP_017828427.1
7 - 8
1110
66
XM_017972939.1
XP_017828428.1
10 - 11
1220
66
XM_017972940.1
XP_017828429.1
10 - 11
1203
66
XM_017972941.1
XP_017828430.1
7 - 8
1284
66
XM_017972942.1
XP_017828431.1
10 - 11
1219
66
XM_017972943.1
XP_017828432.1
10 - 11
1222
66
XM_017972944.1
XP_017828433.1
7 - 8
1388
66
XM_017972945.1
XP_017828434.1
8 - 9
1098
66
XM_017972946.1
XP_017828435.1
8 - 9
1096
66
XM_017972947.1
XP_017828436.1
6 - 7
838
66
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0