This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06104851_m1
See other AP1S2 GE Assays ›
Gene Symbol:
AP1S2
Gene Name:
adaptor related protein complex 1 subunit sigma 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 14001425 - 14047886 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06104851_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AP1S2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008988999.2 XP_008987247.1 1 - 2 121 81
XM_017968369.1 XP_017823858.1 1 - 2 85 81
XM_017968370.1 XP_017823859.1 1 - 2 81 81
XM_017968371.1 XP_017823860.1 1 - 2 85 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017968370.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0