Assay Details
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006339.2 | XP_009004587.1 | 1 - 2 | 286 | 70 | |
XM_017977218.1 | XP_017832707.1 | 1 - 2 | 318 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
