Assay Details
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008979376.1 | XP_008977624.1 | 5 - 6 | 606 | 77 | |
XM_008979377.1 | XP_008977625.1 | 5 - 6 | 624 | 77 | ||
XM_008979378.1 | XP_008977626.1 | 5 - 6 | 611 | 77 | ||
XM_017963800.1 | XP_017819289.1 | 4 - 5 | 706 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
