This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06025204_m1
See other ALPL GE Assays ›
Gene Symbol:
ALPL
Gene Name:
alkaline phosphatase, liver/bone/kidney
Gene Aliases:
-
Chromosome Location:
Chr.7: 55488656 - 55561739 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06025204_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALPL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001185034.1 NP_001171963.1 6 - 7 844 58
XM_009000519.2 XP_008998767.1 6 - 7 911 58
XM_009000520.2 XP_008998768.1 6 - 7 840 58
XM_009000521.2 XP_008998769.1 6 - 7 1328 58
XM_009000522.2 XP_008998770.1 7 - 8 965 58
XM_009000523.1 XP_008998771.1 6 - 7 902 58
XM_009000524.1 XP_008998772.1 4 - 5 487 58
XM_017973935.1 XP_017829424.1 6 - 7 996 58
XM_017973936.1 XP_017829425.1 6 - 7 854 58
GenBank mRNA GQ465850.1 - 6 - 7 844 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0