This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06013214_mH
See other KLRC2 GE Assays ›
Gene Symbol:
KLRC2
Gene Name:
killer cell lectin like receptor C2
Gene Aliases:
NKG2CE
Chromosome Location:
Chr.9: 10862732 - 10870025 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06013214_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KLRC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257247.1 NP_001244176.1 4 - 5 516 91
XM_009003263.2 XP_009001511.1 5 - 6 633 91
XM_017975765.1 XP_017831254.1 5 - 6 649 91
XM_017975766.1 XP_017831255.1 5 - 6 649 91
XM_017975767.1 XP_017831256.1 5 - 6 649 91
XM_017975768.1 XP_017831257.1 5 - 6 649 91
XM_017975770.1 XP_017831259.1 5 - 6 649 91
XM_017975771.1 XP_017831260.1 5 - 6 647 91
XM_017975772.1 XP_017831261.1 5 - 6 644 91
GenBank mRNA EF050434.1 - 4 - 5 516 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0