Assay Details
Gene Symbol: SLC16A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752674.4 | XP_002752720.1 | 4 - 5 | 799 | 72 | |
XM_009004130.2 | XP_009002378.1 | 4 - 5 | 1293 | 72 | ||
XM_009004131.2 | XP_009002379.1 | 4 - 5 | 798 | 72 | ||
XM_009004132.2 | XP_009002380.1 | 4 - 5 | 816 | 72 | ||
XM_009004133.2 | XP_009002381.1 | 4 - 5 | 1309 | 72 | ||
XM_009004134.2 | XP_009002382.1 | 3 - 4 | 513 | 72 | ||
XM_009004135.2 | XP_009002383.1 | 3 - 4 | 403 | 72 | ||
XM_017976172.1 | XP_017831661.1 | 5 - 6 | 1401 | 72 | ||
XM_017976173.1 | XP_017831662.1 | 5 - 6 | 997 | 72 | ||
XM_017976174.1 | XP_017831663.1 | 5 - 6 | 909 | 72 | ||
XM_017976175.1 | XP_017831664.1 | 3 - 4 | 2242 | 72 | ||
XM_017976176.1 | XP_017831665.1 | 3 - 4 | 1020 | 72 | ||
XM_017976177.1 | XP_017831666.1 | 4 - 5 | 799 | 72 | ||
XM_017976178.1 | XP_017831667.1 | 4 - 5 | 799 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
