Assay Details
Gene Symbol: LOC100402032 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754365.3 | XP_002754411.1 | 11 - 12 | 1123 | 102 | |
XM_017977518.1 | XP_017833007.1 | 11 - 12 | 994 | 102 |

More Information
Set Membership: |
![]() ![]() |
