This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05975739_m1
See other LOC100403511 GE Assays ›
Gene Symbol:
LOC100403511
Gene Name:
Golgi pH regulator
Gene Aliases:
GPR89A, GPR89B
Chromosome Location:
Chr.18: 1512086 - 1579416 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05975739_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100403511
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002759822.4 XP_002759868.1 7 - 8 785 81
XM_008984197.2 XP_008982445.1 7 - 8 785 81
XM_008984200.1 XP_008982448.1 6 - 7 677 81
XM_017966197.1 XP_017821686.1 5 - 6 539 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0