This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05974104_m1
See other LOC108593534 GE Assays ›
Gene Symbol:
LOC108593534
Gene Name:
tubulin polyglutamylase TTLL5-like
Gene Aliases:
-
Chromosome Location:
-
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05974104_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC108593534
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017977232.1 XP_017832721.1 2 - 3 735 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Panther Classification:

Molecular Function -

microtubule or microtubule-binding cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0