This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05973043_m1
See other PHACTR2 GE Assays ›
Gene Symbol:
PHACTR2
Gene Name:
phosphatase and actin regulator 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 144297505 - 144511141 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05973043_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002747098.4 XP_002747144.1 8 - 9 1792 62
XM_008995182.2 XP_008993430.1 8 - 9 4962 62
XM_008995183.2 XP_008993431.1 7 - 8 1549 62
XM_008995184.2 XP_008993432.1 7 - 8 4716 62
XM_017971447.1 XP_017826936.1 7 - 8 1625 62
XM_017971448.1 XP_017826937.1 7 - 8 1549 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0