Assay Details
Gene Symbol: OPALIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009010081.1 | XP_009008329.1 | 3 - 4 | 512 | 83 | |
XM_017979196.1 | XP_017834685.1 | 3 - 4 | 3200 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017979196.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
