This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05972801_m1
See other MRPL55 GE Assays ›
Gene Symbol:
MRPL55
Gene Name:
mitochondrial ribosomal protein L55
Gene Aliases:
-
Chromosome Location:
Chr.19: 33488083 - 33491413 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05972801_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002760834.3 XP_002760880.1 3 - 4 437 69
XM_008985663.2 XP_008983911.1 4 - 5 492 69
XM_008985664.2 XP_008983912.1 4 - 5 519 69
XM_008985665.2 XP_008983913.1 4 - 5 510 69
XM_008985666.2 XP_008983914.1 4 - 5 477 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0