Assay Details
Gene Symbol: TMEM132A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755384.2 | XP_002755430.1 | 4 - 5 | 1040 | 55 | |
XM_017978177.1 | XP_017833666.1 | 4 - 5 | 1236 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
